Läs mer
4-Ethylpyrimidine-5-carboxylic Acid, TRC
Produktkod. 30333974
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30333974 100 mg
30331691 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30333974 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB436693100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Ethylpyrimidine-5-carboxylic Acid
CAS 1147710-24-4
Molekylformel C7H8N2O2
Kvantitet 100 mg
InChI formel InChI=1S/C7H8N2O2/c1-2-6-5(7(10)11)3-8-4-9-6/h3-4H,2H2,1H3,(H,10,11)
LEDER CCc1ncncc1C(=O)O
IUPAC-namn 4-ethylpyrimidine-5-carboxylic acid
Molekylvikt (g/mol) 152.15
Formel vikt 152.0586