missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,3-dioxolan-2-one, 98%, Thermo Scientific Chemicals

Catalog Number p-7053812
Click to view available options
Quantity:
25 g
100 g
114435-02-8
C3H3FO3
106.052
MFCD06247543
SBLRHMKNNHXPHG-UHFFFAOYSA-N
fluoroethylene carbonate, 1,3-dioxolan-2-one, 4-fluoro, fluoro ethylene carbonate, 4-fluoro-1,3-dioxalan-2-one, 4-fluoro-2-oxo-1,3-dioxolane, 4-fluoro-ethylene carbonate, 4-fluoro-ethylene carboante, ksc219e1h, fec, lithium ion battery grade
2769656
4-fluoro-1,3-dioxolan-2-one
C1C(OC(=O)O1)F
114435-02-8
C3H3FO3
106.052
MFCD06247543
SBLRHMKNNHXPHG-UHFFFAOYSA-N
fluoroethylene carbonate, 1,3-dioxolan-2-one, 4-fluoro, fluoro ethylene carbonate, 4-fluoro-1,3-dioxalan-2-one, 4-fluoro-2-oxo-1,3-dioxolane, 4-fluoro-ethylene carbonate, 4-fluoro-ethylene carboante, ksc219e1h, fec, lithium ion battery grade
2769656
4-fluoro-1,3-dioxolan-2-one
C1C(OC(=O)O1)F

4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 18°C to 23°C
Color Colorless
Boiling Point 212°C
Flash Point >102°C (216°F)
Quantity 25 g
Solubility Information Slightly miscible with water.
Formula Weight 106.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-1,3-dioxolan-2-one
compliance-icons
Product Identifier
  • 4-Fluoro-1,3-dioxolan-2-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Fluoro-1,3-dioxolan-2-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'