Läs mer
4-Fluoro-2-(hydroxymethyl)phenol, TRC
Produktkod. 30137737
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137737 500 mg
30135786 1 g
17932515 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137737 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF532030500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2357-33-7
Molekylformel C7 H7 F O2
Molekylvikt (g/mol) 142.128
Synonym Tavaborole Impurity B, 5-Fluoro-2-hydroxybenzenemethanol, 5-Fluoro-2-hydroxybenzyl Alcohol, Benzenemethanol, 5-fluoro-2-hydroxy-
IUPAC-namn 4-fluoro-2-(hydroxymethyl)phenol
LEDER OCc1cc(F)ccc1O
Specifikationer
Kemiskt namn eller material 4-Fluoro-2-(hydroxymethyl)phenol
Kvantitet 500 mg
InChI formel InChI=1S/C7H7FO2/c8-6-1-2-7(10)5(3-6)4-9/h1-3,9-10H,4H2
Formel vikt 142.043