Learn More
4-Fluoro-2-methyl(thiobenzamide), 97%, Thermo Scientific™
Product Code. 15486027
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1256482-73-1
Molecular Formula C8H8FNS
Molecular Weight (g/mol) 169.217
MDL Number MFCD09757564
InChI Key CIVOYVGIXSEIKD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methyl-thiobenzamide, 4-fluoro-2-methyl thiobenzamide, 4-fluoro-2-methylbenzene-1-carbothioamide
PubChem CID 46779082
IUPAC Name 4-fluoro-2-methylbenzenecarbothioamide
SMILES CC1=C(C=CC(=C1)F)C(=S)N
Specifications
Melting Point 52°C to 56°C
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methyl(thiobenzamide)