Läs mer
4-Fluoro-2-methylphenylacetic acid, 97%, Thermo Scientific™
Produktkod. p-7028948
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15443457 250 mg 250mg
11491280 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15443457 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26365.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Fluoro-2-methylphenylacetic acid is used as a synthetic building block.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 407640-40-8
Molekylformel C9H9FO2
Molekylvikt (g/mol) 168.167
MDL-nummer MFCD00049318
InChI-nyckel KOZXQTAPRQFRLT-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylphenylacetic acid, 2-4-fluoro-2-methylphenyl acetic acid, 4-fluoro-2-methylphenyl acetic acid, 4-fluoro-2-methyl-phenyl-acetic acid, benzeneacetic acid, 4-fluoro-2-methyl, pubchem4174, 2-4-fluoro-2-methyl-phenyl acetic acid, acmc-1akub, timtec-bb sbb027289, 4-fluoro-2-methylphenylaceticacid
PubChem CID 5016950
IUPAC-namn 2-(4-fluoro-2-methylphenyl)acetic acid
LEDER CC1=C(C=CC(=C1)F)CC(=O)O
Specifikationer
Kemiskt namn eller material 4-Fluoro-2-methylphenylacetic acid
Smältpunkt 96°C to 99°C
Kvantitet 250 mg
Löslighetsinformation Insoluble in water.
Formel vikt 168.17
Procent renhet 97%