Learn More
4-Fluoro-3-nitroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Insoluble in water.
Notes
Incompatible with Oxidizing agents, Acids, Acid chlorides, Acid anhydrides.
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Insoluble in water.
Notes
Incompatible with Oxidizing agents, Acids, Acid chlorides, Acid anhydrides.
Specifications
Melting Point | 94°C to 98°C |
Flash Point | 91°C (195°F) |
Quantity | 25 g |
Beilstein | 2210199 |
Solubility Information | Insoluble in water. |
Formula Weight | 156.12 |
Percent Purity | 98% |
Chemical Name or Material | 4-Fluoro-3-nitroaniline |