missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-3-phenoxybenzaldehyde, 97%

Brand:  Alfa Aesar™ H61202.06

Additional Details : CAS Number : 68359-57-9 Weight : 0.00500kg Transport : UN number : 3082 Chem class : 9 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15437708

  • 529.88 SEK / 5g


Estimated Shipment
30-10-2020
Add to basket

Specifications

Chemical Identifiers

68359-57-9
216.211
JDICMOLUAHZVDS-UHFFFAOYSA-N
110068
C1=CC=C(C=C1)OC2=C(C=CC(=C2)C=O)F
C13H9FO2
MFCD01318148
4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
4-fluoro-3-phenoxybenzaldehyde

Specifications

4-Fluoro-3-phenoxybenzaldehyde
>110°C (230°F)
Colorless
68359-57-9
MFCD01318148
4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
C1=CC=C(C=C1)OC2=C(C=CC(=C2)C=O)F
216.211
216.21
97%
1.209 g/mL
1.583
5g
C13H9FO2
UN3082
JDICMOLUAHZVDS-UHFFFAOYSA-N
4-fluoro-3-phenoxybenzaldehyde
110068
Liquid
Videos
SDS Documents