Läs mer
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid, 98%
Produktkod. p-7034784
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15439835 1 g 1g
15449835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15439835 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H52483.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 182344-23-6 | C7H5BF4O2 | 207.919 g/mol
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methabnol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 182344-23-6
Molekylformel C7H5BF4O2
Molekylvikt (g/mol) 207.919
MDL-nummer MFCD07363787
InChI-nyckel GUJYFCBXDUPORN-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl phenylboronic acid, 4-fluoro-3-trifluoromethylphenylboronic acid, 4-fluoro-3-trifluoromethyl benzeneboronic acid, 5-borono-2-fluorobenzotrifluoride, 4-fluoro-3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl-4-fluoro-phenylboronic acid, boronic acid, 4-fluoro-3-trifluoromethyl phenyl, 4-fluoro-3-trifluoromethyl benzeneboronicacid
PubChem CID 17750051
IUPAC-namn [4-fluoro-3-(trifluoromethyl)phenyl]boronic acid
LEDER B(C1=CC(=C(C=C1)F)C(F)(F)F)(O)O
Spécification
Kemiskt namn eller material 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid
Smältpunkt 170°C to 174°C
Kvantitet 1 g
Löslighetsinformation Soluble in methabnol.
Formel vikt 207.92
Procent renhet 98%
RUO – Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.