Learn More
Alfa Aesar™ 4-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-3-(trifluoromethyl)benzamide, 95%
Brand:  Alfa Aesar™ H65558.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15459736
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 02-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-3-(trifluoromethyl)benzamide
337.32
MFCD27975784
Specifications
4-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-3-(trifluoromethyl)benzamide
Solid
337.32
1g
95%
MFCD27975784