missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluorobiphenyl-4-boronic acid, 98%, Thermo Scientific™

Catalog Number p-7055762
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
140369-67-1
C12H10BFO2
216.02
MFCD08544387
KHMFYFVXTICBEL-UHFFFAOYSA-N
4-4-fluorophenyl phenylboronic acid, 4'-fluoro-1,1'-biphenyl-4-yl boronic acid, 4'-fluoro-4-biphenylboronic acid, 4'-fluorobiphenyl-4-ylboronic acid, 4-4-fluorophenyl phenylboronicacid, 4'-fluorobiphenyl-4-boronic acid, boronic acid,b-4'-fluoro 1,1'-biphenyl-4-yl, 4'-fluoro-1,1'-biphenyl-4-ylboronic acid, akos brn-1038, pubchem11575
14985212
[4-(4-fluorophenyl)phenyl]boronic acid
OB(O)C1=CC=C(C=C1)C1=CC=C(F)C=C1
This item is not returnable. View return policy
140369-67-1
C12H10BFO2
216.02
MFCD08544387
KHMFYFVXTICBEL-UHFFFAOYSA-N
4-4-fluorophenyl phenylboronic acid, 4'-fluoro-1,1'-biphenyl-4-yl boronic acid, 4'-fluoro-4-biphenylboronic acid, 4'-fluorobiphenyl-4-ylboronic acid, 4-4-fluorophenyl phenylboronicacid, 4'-fluorobiphenyl-4-boronic acid, boronic acid,b-4'-fluoro 1,1'-biphenyl-4-yl, 4'-fluoro-1,1'-biphenyl-4-ylboronic acid, akos brn-1038, pubchem11575
14985212
[4-(4-fluorophenyl)phenyl]boronic acid
OB(O)C1=CC=C(C=C1)C1=CC=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 8689987
Formula Weight 216.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4'-Fluorobiphenyl-4-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'