Läs mer
4'-Fluorobutyrophenone, 97%, Thermo Scientific™
Produktkod. p-7028640
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
Produktkod. 11391959
Brand: Thermo Scientific Alfa Aesar H26034.03
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4′-fluorobutyrophenone derivatives is used in the preparation of its derivatives that includes haloperidol, haloanisone, triperidol, methylperidide, haloperidide and dipiperone.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 582-83-2
Molekylformel C10H11FO
Molekylvikt (g/mol) 166.195
MDL-nummer MFCD00017967
InChI-nyckel QHDXPJMOWRLLRV-UHFFFAOYSA-N
PubChem CID 68498
IUPAC-namn 1-(4-fluorophenyl)butan-1-one
LEDER CCCC(=O)C1=CC=C(C=C1)F
Specifikationer
Kemiskt namn eller material 4'-Fluorobutyrophenone
Smältpunkt 35°C to 38°C
Kvantitet 1 g
Synonym 4'-fluorobutyrophenone, 1-4-fluorophenyl butan-1-one, butyrophenone, 4'-fluoro, p-fluorobutyrophenone, 1-4-fluoro-phenyl-butan-1-one, 1-butanone, 1-4-fluorophenyl, 1-4-fluorophenyl-1-butanone, acmc-20apap, 4-fluorobutyrophenone, 1-butanone,1-4-fluorophenyl
Löslighetsinformation Insoluble in water.
Formel vikt 166.2
Procent renhet 97%