missing translation for 'onlineSavingsMsg'
Läs mer

4-Fluorophenylacetyl chloride, 97%

Produktkod. p-7047438
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11304106 1 g 1g
11314106 5 g 5g
11324106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11304106 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L19235.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 459-04-1 | C8H6ClFO | 172.58 g/mol

4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

Solubility
Miscible with dichloromethane.

Notes
Moisture Sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 459-04-1
Molekylformel C8H6ClFO
Molekylvikt (g/mol) 172.58
MDL-nummer MFCD00674181
InChI-nyckel SIOJFYRPBYGHOO-UHFFFAOYSA-N
Synonym 4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
PubChem CID 3744988
IUPAC-namn 2-(4-fluorophenyl)acetyl chloride
LEDER FC1=CC=C(CC(Cl)=O)C=C1

Tekniske data

Kemiskt namn eller material 4-Fluorophenylacetyl chloride
Densitet 1.259
Kokpunkt 60°C (8 mmHg)
Flampunkt 107°C (224°F)
Brytningsindex 1.512
Kvantitet 1 g
UN-nummer UN3265
Beilstein 972148
Känslighet Moisture sensitive
Löslighetsinformation Miscible with dichloromethane.
Formel vikt 172.59
Procent renhet 97%
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.