Learn More
4-Fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-3778637
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
1765-93-1
C6H6BFO2
139.92
MFCD00039136
LBUNNMJLXWQQBY-UHFFFAOYSA-N
4-fluorobenzeneboronic acid, 4-fluorophenyl boronic acid, 4-fluorophenyl boranediol, p-fluorophenylboronic acid, 4-fluorophenylbornic acid, 4-fluorophenyl dihydroxyboron, 4-fluorophenyl dihydroxyborane, p-fluorophenyl boric acid, 4-fluorophenyl boric acid
285645
CHEBI:48661
(4-fluorophenyl)boronic acid
OB(O)C1=CC=C(F)C=C1
1765-93-1
C6H6BFO2
139.92
MFCD00039136
LBUNNMJLXWQQBY-UHFFFAOYSA-N
4-fluorobenzeneboronic acid, 4-fluorophenyl boronic acid, 4-fluorophenyl boranediol, p-fluorophenylboronic acid, 4-fluorophenylbornic acid, 4-fluorophenyl dihydroxyboron, 4-fluorophenyl dihydroxyborane, p-fluorophenyl boric acid, 4-fluorophenyl boric acid
285645
CHEBI:48661
(4-fluorophenyl)boronic acid
OB(O)C1=CC=C(F)C=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 255.0°C to 259.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 139.92
Percent Purity 97%
Physical Form Crystalline or Fluffy Powder
Chemical Name or Material 4-Fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.