missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3778629
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
823-85-8
C6H8ClFN2
162.59
MFCD00012942
FEKUXLUOKFSMRO-UHFFFAOYSA-N
4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride
69981
[H+].[Cl-].NNC1=CC=C(F)C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
823-85-8
C6H8ClFN2
162.59
MFCD00012942
FEKUXLUOKFSMRO-UHFFFAOYSA-N
4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride
69981
[H+].[Cl-].NNC1=CC=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 275°C to 277°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula FC6H4NHNH2·HCl
Quantity 10 g
Beilstein 15, I, 105
Solubility Information Solubility in water: soluble
IUPAC Name (4-fluorophenyl)hydrazine;hydrochloride
Formula Weight 162.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Fluorophenylhydrazine hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Fluorophenylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'