Läs mer
4-Hexylpyridine, TRC
Produktkod. 30403383
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30403383 100 mg
30399151 500 mg
30395638 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30403383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH324960100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27876-24-0
Molekylformel C11 H17 N
Molekylvikt (g/mol) 163.259
IUPAC-namn 4-hexylpyridine
LEDER CCCCCCc1ccncc1
Specifikationer
Kemiskt namn eller material 4-Hexylpyridine
Kvantitet 100 mg
InChI formel InChI=1S/C11H17N/c1-2-3-4-5-6-11-7-9-12-10-8-11/h7-10H,2-6H2,1H3
Formel vikt 163.136