Läs mer
4-HO-DPHP, TRC
Produktkod. 30404363
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30404363 10 mg
1 options
Produktkod. 30404363 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH45642010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-HO-DPHP
CAS 114527-61-6
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H11 O5 P
Kvantitet 10 mg
Synonym 4-Hydroxyphenyl Phenyl Hydrogen Phosphate, 4-Hydroxyphenyl Diphenyl Phosphate, Phosphoric Acid (4-Hydroxyphenyl)diphenyl Ester
InChI formel InChI=1S/C12H11O5P/c13-10-6-8-12(9-7-10)17-18(14,15)16-11-4-2-1-3-5-11/h1-9,13H,(H,14,15)
LEDER Oc1ccc(OP(=O)(O)Oc2ccccc2)cc1
IUPAC-namn (4-hydroxyphenyl) phenyl hydrogen phosphate
Molekylvikt (g/mol) 266.187
Formel vikt 266.034