Läs mer
4-Hydroxy-1-(3-pyridyl)-1-butanone, TRC
Produktkod. 30011319
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011319 5 mg
30010223 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011319 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9530005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59578-62-0
Molekylformel C9 H11 N O2
Molekylvikt (g/mol) 165.19
Synonym 4-Hydroxy-1-(3-pyridinyl)-1-butanone, 4-Hydroxy-1-(3-pyridyl)-1-butanone, 4-Oxo-4-(3-pyridyl)-1-butanol
IUPAC-namn 4-hydroxy-1-pyridin-3-ylbutan-1-one
LEDER OCCCC(=O)c1cccnc1
Specifikationer
Kemiskt namn eller material 4-Hydroxy-1-(3-pyridyl)-1-butanone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C9H11NO2/c11-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,11H,2,4,6H2
Formel vikt 165.079
Procent renhet >95%