missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-4509649
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
56-04-2
C5H6N2OS
142.18
MFCD00006040
HWGBHCRJGXAGEU-UHFFFAOYSA-N
methylthiouracil, 6-methyl-2-thiouracil, methiocil, 4-hydroxy-2-mercapto-6-methylpyrimidine, thimecil, thiomecil, thiothymin, alkiron, metacil, 2-mercapto-6-methylpyrimidin-4-ol
667493
CHEBI:82346
6-methyl-2-sulfanylidene-1H-pyrimidin-4-one
CC1=CC(=O)NC(=S)N1
This item is not returnable. View return policy
56-04-2
C5H6N2OS
142.18
MFCD00006040
HWGBHCRJGXAGEU-UHFFFAOYSA-N
methylthiouracil, 6-methyl-2-thiouracil, methiocil, 4-hydroxy-2-mercapto-6-methylpyrimidine, thimecil, thiomecil, thiothymin, alkiron, metacil, 2-mercapto-6-methylpyrimidin-4-ol
667493
CHEBI:82346
6-methyl-2-sulfanylidene-1H-pyrimidin-4-one
CC1=CC(=O)NC(=S)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 330.0°C
Color Colorless to Yellow
Loss on Drying 0.5% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 24,III,1289
Merck Index 15,6201
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in alcohols,acetone,ether,nh3,insoluble in benzene,chloroform
Formula Weight 142.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-2-mercapto-6-methylpyrimidine
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 4-Hydroxy-2-mercapto-6-methylpyrimidine
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Indicaciones de peligro
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'