missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-4258996
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18278-34-7
C8H8O3
152.15
MFCD00051964
WBIZZNFQJPOKDK-UHFFFAOYSA-N
4-hydroxy-o-anisaldehyde, benzaldehyde, 4-hydroxy-2-methoxy, 4-hydroxyl-2-methoxyl benzaldehyde, 4-formyl-3-methoxyphenol, 2-formyl-5-hydroxyanisole, 4-hydroxy-2-methoxybenaldehyde, 4-hydroxy-2-methoxybenzaldehdye, pubchem8130, acmc-209eju, argogel-mb-cho
519541
4-hydroxy-2-methoxybenzaldehyde
COC1=CC(O)=CC=C1C=O
This item is not returnable. View return policy
18278-34-7
C8H8O3
152.15
MFCD00051964
WBIZZNFQJPOKDK-UHFFFAOYSA-N
4-hydroxy-o-anisaldehyde, benzaldehyde, 4-hydroxy-2-methoxy, 4-hydroxyl-2-methoxyl benzaldehyde, 4-formyl-3-methoxyphenol, 2-formyl-5-hydroxyanisole, 4-hydroxy-2-methoxybenaldehyde, 4-hydroxy-2-methoxybenzaldehdye, pubchem8130, acmc-209eju, argogel-mb-cho
519541
4-hydroxy-2-methoxybenzaldehyde
COC1=CC(O)=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 153.0°C to 158.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH(O)C6H3(OH)OCH3
Quantity 1 g
Solubility Information Solubility in water: practically insoluble in water (20°C)
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-2-methoxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxy-2-methoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'