Läs mer
4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Produktkod. 30463078
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30463078 4 x 25 mg
17962236 25 mg
17961206 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30463078 Leverantör Mikromol Leverantörsnummer MM0084.0900254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 24683-25-8
Analyt- eller komponentnamn 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Frakt skick Room Temperature
Molekylformel C10 H10 N2 O4 S
Certifikat och efterlevnad ISO 9001
Synonym 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Piroxicam Imp. C (EP), Piroxicam Impurity C
InChI formel InChI=1S/C10H10N2O4S/c1-12-8(10(11)14)9(13)6-4-2-3-5-7(6)17(12,15)16/h2-5,13H,1H3,(H2,11,14)
Lösningstyp Neat
LEDER O=C(N)C1=C(O)C=2C=CC=CC2S(=O)(=O)N1C
IUPAC-namn 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvikt (g/mol) 254.26
Kvantitet 4 x 25 mg
Formel vikt 254.0361 g/mol