missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methylquinoline, 98+%, Thermo Scientific Chemicals

Catalog Number 11461998
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
607-67-0
C10H9NO
159.19
MFCD00006758,MFCD00518775
NWINIEGDLHHNLH-UHFFFAOYSA-N
2-methylquinolin-4-ol, 4-hydroxy-2-methylquinoline, 2-methyl-4-quinolinol, 4-quinolinol, 2-methyl, 4-hydroxyquinaldine, 2-methyl-4-hydroxyquinoline, 2-methyl-quinolin-4-ol, 2-methylquinolin-4 1h-one, 2-methyl-4 1h-quinolinone, 2-methyl-1,4-dihydroquinolin-4-one
69089
CC1=CC(=O)C2=CC=CC=C2N1
This item is not returnable. View return policy
607-67-0
C10H9NO
159.19
MFCD00006758,MFCD00518775
NWINIEGDLHHNLH-UHFFFAOYSA-N
2-methylquinolin-4-ol, 4-hydroxy-2-methylquinoline, 2-methyl-4-quinolinol, 4-quinolinol, 2-methyl, 4-hydroxyquinaldine, 2-methyl-4-hydroxyquinoline, 2-methyl-quinolin-4-ol, 2-methylquinolin-4 1h-one, 2-methyl-4 1h-quinolinone, 2-methyl-1,4-dihydroquinolin-4-one
69089
CC1=CC(=O)C2=CC=CC=C2N1

4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 234°C to 236°C
Quantity 5 g
Beilstein 118320
Solubility Information Slightly soluble in water.
IUPAC Name 2-methyl-1H-quinolin-4-one
Formula Weight 159.19
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-2-methylquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'