Läs mer
4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone, Mikromol™
Produktkod. 30052621
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052621 25 mg
30050815 100 mg
30460228 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30052621 Leverantör Mikromol Leverantörsnummer MM3329.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone
CAS 134612-80-9
Analyt- eller komponentnamn 4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone
Frakt skick Room Temperature
Molekylformel C15 H13 N O5
Certifikat och efterlevnad ISO 17025
Synonym Methanone, (4-hydroxy-3-methoxy-5-nitrophenyl)(4-methylphenyl)-, 4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone, Ro 40-7591, Tolcapone Related Compound B
InChI formel InChI=1S/C15H13NO5/c1-9-3-5-10(6-4-9)14(17)11-7-12(16(19)20)15(18)13(8-11)21-2/h3-8,18H,1-2H3
LEDER COc1cc(cc(c1O)[N+](=O)[O-])C(=O)c2ccc(C)cc2
IUPAC-namn (4-hydroxy-3-methoxy-5-nitrophenyl)-(4-methylphenyl)methanone
Molekylvikt (g/mol) 287.27
Kvantitet 25 mg
Formel vikt 287.0794 g/mol