missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7025248
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7149-10-2
MFCD00012864
This item is not returnable. View return policy
7149-10-2
MFCD00012864

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼220°C (decomposition)
Molecular Formula C8H11ClNO2
Quantity 1 g
Beilstein 3915418
Sensitivity Moisture sensitive
Synonym 4-hydroxy-3-methoxybenzylamine hydrochloride, 4-aminomethyl-2-methoxyphenol hydrochloride, vanillylamine hydrochloride, n-vanillylamine hydrochloride, 2-vanillylamine hydrochloride, 4-hydroxy-3-methoxybenzylamine hcl, phenol, 4-aminomethyl-2-methoxy-, hydrochloride, 4-aminomethyl-2-methoxyphenol, chloride, vanillylamine hcl
InChI Key LOYPVODLNGWOLM-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)CN)O.Cl
IUPAC Name 4-(aminomethyl)-2-methoxyphenol;hydrochloride
Molecular Weight (g/mol) 188.63
PubChem CID 165576
Formula Weight 189.64
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-methoxybenzylamine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'