missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-nitrostilbene, 97%, Thermo Scientific™

Catalog Number p-7077354
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
19221-08-0
C14H11NO3
241.25
OETQWIHJPIESQB-OWOJBTEDSA-N
4-hydroxy-4'-nitrostilbene, 4-4-nitrostyryl phenol, 4-e-2-4-nitrophenyl ethenyl phenol, 4-hydroxy-4/'-nitrostilbene, 4-1e-2-4-nitrophenyl vinyl phenol, 4'-nitrostilben-4-ol, cambridge id 5309411, e-4'-nitrostilbene-4-ol, e-4-4-nitrostyryl phenol
759250
4-[(E)-2-(4-nitrophenyl)ethenyl]phenol
C1=CC(=CC=C1C=CC2=CC=C(C=C2)O)[N+](=O)[O-]
This item is not returnable. View return policy
19221-08-0
C14H11NO3
241.25
OETQWIHJPIESQB-OWOJBTEDSA-N
4-hydroxy-4'-nitrostilbene, 4-4-nitrostyryl phenol, 4-e-2-4-nitrophenyl ethenyl phenol, 4-hydroxy-4/'-nitrostilbene, 4-1e-2-4-nitrophenyl vinyl phenol, 4'-nitrostilben-4-ol, cambridge id 5309411, e-4'-nitrostilbene-4-ol, e-4-4-nitrostyryl phenol
759250
4-[(E)-2-(4-nitrophenyl)ethenyl]phenol
C1=CC(=CC=C1C=CC2=CC=C(C=C2)O)[N+](=O)[O-]
Melting Point 209.0°C to 210.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 241.25
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-4'-nitrostilbene
compliance-icons
Product Identifier
  • 4-Hydroxy-4'-nitrostilbene, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'