Läs mer
4-Hydroxy-7-methoxy-6-quinolinecarboxamide, TRC
Produktkod. 30395847
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30395847 50 mg
30396537 10 mg
30392793 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30395847 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH21104050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2110414-05-4
Molekylformel C11 H10 N2 O3
Molekylvikt (g/mol) 218.209
Synonym 4-Hydroxy-7-Methoxyquinoline-6-Carboxamide
IUPAC-namn 4-hydroxy-7-methoxyquinoline-6-carboxamide
LEDER COc1cc2nccc(O)c2cc1C(=O)N
Specifikationer
Kemiskt namn eller material 4-Hydroxy-7-methoxy-6-quinolinecarboxamide
Kvantitet 50 mg
InChI formel InChI=1S/C11H10N2O3/c1-16-10-5-8-6(4-7(10)11(12)15)9(14)2-3-13-8/h2-5H,1H3,(H2,12,15)(H,13,14)
Formel vikt 218.069