Läs mer
4-Hydroxy Atorvastatin Disodium Salt, TRC
Produktkod. 30007392
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007392 5 mg
30011167 10 mg
30009598 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007392 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8289605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1276537-18-8
Molekylformel C33H33FN2Na2O6
Molekylvikt (g/mol) 618.6
LEDER [Na+].[Na+].CC(C)c1c(C(=O)Nc2ccc([O-])cc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)[O-]
Specifikationer
Kemiskt namn eller material p-Hydroxy Atorvastatin, Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C33H35FN2O6/c1-20(2)31-30(33(42)35-24-12-14-25(37)15-13-24)29(21-6-4-3-5-7-21)32(22-8-10-23(34)11-9-22)36(31)17-16-26(38)18-27(39)19-28(40)41/h3-15,20,26-27,37-39H,16-19H2,1-2H3,(H,35,42)(H,40,41)/t26-,27-/m1/s1
Procent renhet >95%