Läs mer
4-Hydroxy-benzenebutanoic Acid Methyl Ester, TRC
Produktkod. 30344246
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30344246 500 mg
1 options
Produktkod. 30344246 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB185298500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxy-benzenebutanoic Acid Methyl Ester
CAS 22320-10-1
Molekylformel C11H14O3
Kvantitet 500 mg
Synonym 4-(p-Hydroxyphenyl)butyric Acid Methyl Ester, Methyl 4-(4-Hydroxyphenyl)butanoate, Methyl 4-(4-Hydroxyphenyl)butyrate, Methyl Ester 4-Hydroxy Benzene butanoic Acid
InChI formel InChI=1S/C11H14O3/c1-14-11(13)4-2-3-9-5-7-10(12)8-6-9/h5-8,12H,2-4H2,1H3
LEDER COC(=O)CCCc1ccc(O)cc1
IUPAC-namn methyl 4-(4-hydroxyphenyl)butanoate
Molekylvikt (g/mol) 194.23
Formel vikt 194.0943