Läs mer
4-Hydroxy-clomiphene, TRC
Produktkod. 30003620
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003620 10 mg
30011964 1 mg
30006753 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH91840510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79838-51-0
Molekylformel C26 H28 Cl N O2
Molekylvikt (g/mol) 421.96
Synonym 4-[2-Chloro-1-[4-[2-(diethylamino)ethoxy]phenyl]-2-phenylethenyl]phenol
IUPAC-namn 4-[2-chloro-1-[4-[2-(diethylamino)ethoxy]phenyl]-2-phenyl-vinyl]phenol
LEDER CCN(CC)CCOc1ccc(cc1)C(=C(Cl)c2ccccc2)c3ccc(O)cc3
Specifikationer
Kemiskt namn eller material 4-Hydroxy-clomiphene
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C26H28ClNO2/c1-3-28(4-2)18-19-30-24-16-12-21(13-17-24)25(20-10-14-23(29)15-11-20)26(27)22-8-6-5-7-9-22/h5-17,29H,3-4,18-19H2,1-2H3
Formel vikt 421.1809
Procent renhet >95%