missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-N-methylbutanamide, TRC

Product Code. 17961185
Change view
Click to view available options
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
17961185 1 g
17962135 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17961185 Supplier Toronto Research Chemicals Supplier No. TRCH9438351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37941-69-8
Molekylformel C5 H11 N O2
Molekylvikt (g/mol) 117.15
Synonym N-Methyl-γ-hydroxybutyramide, 4-Hydroxy-N-methylbutanamide
IUPAC-namn 4-hydroxy-N-methyl-butanamide
LEDER CNC(=O)CCCO

Specifications

Kemiskt namn eller material 4-Hydroxy-N-methylbutanamide
Kvantitet 1 g
InChI formel InChI=1S/C5H11NO2/c1-6-5(8)3-2-4-7/h7H,2-4H2,1H3,(H,6,8)
Formel vikt 117.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.