Läs mer
4-Hydroxy-N-methylbutanamide, TRC
Produktkod. 17962135
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962135 10 g
17961185 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17962135 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94383510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37941-69-8
Molekylformel C5 H11 N O2
Molekylvikt (g/mol) 117.15
Synonym N-Methyl-γ-hydroxybutyramide, 4-Hydroxy-N-methylbutanamide
IUPAC-namn 4-hydroxy-N-methyl-butanamide
LEDER CNC(=O)CCCO
Specifikationer
Kemiskt namn eller material 4-Hydroxy-N-methylbutanamide
Kvantitet 10 g
InChI formel InChI=1S/C5H11NO2/c1-6-5(8)3-2-4-7/h7H,2-4H2,1H3,(H,6,8)
Formel vikt 117.079