Läs mer
4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Produktkod. 18063050
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18063050 100 mg
18012250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18063050 Leverantör Mikromol Leverantörsnummer MM0084.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 65897-46-3
Analyt- eller komponentnamn 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Frakt skick Room Temperature
Molekylformel C14 H11 N3 O4 S
Certifikat och efterlevnad ISO 17025
Synonym 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Piroxicam Imp. B (EP), Piroxicam Impurity B
InChI formel InChI=1S/C14H11N3O4S/c18-13-9-5-1-2-6-10(9)22(20,21)17-12(13)14(19)16-11-7-3-4-8-15-11/h1-8,17-18H,(H,15,16,19)
LEDER OC1=C(NS(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3
IUPAC-namn 4-hydroxy-1,1-dioxo-N-pyridin-2-yl-2H-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvikt (g/mol) 317.32
Kvantitet 100 mg
Formel vikt 317.047 g/mol