Läs mer
4'-Hydroxy-PhIP, TRC
Produktkod. 17930575
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17930575 5 mg
17940795 1 mg
17901395 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17930575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9507605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126861-72-1
Molekylformel C13 H12 N4 O
Molekylvikt (g/mol) 240.2606
Synonym 4-(2-Amino-1-methyl-1H-imidazo[4,5-b]pyridin-6-yl)phenol, 2-Amino-4'-hydroxy-1-methyl-6-phenylimidazo[4,5-b]pyridine
IUPAC-namn 4-(2-amino-1-methylimidazo[4,5-b]pyridin-6-yl)phenol
LEDER Cn1c(N)nc2ncc(cc12)c3ccc(O)cc3
Specifikationer
Kemiskt namn eller material 4'-Hydroxy-PhIP
Kvantitet 5 mg
InChI formel InChI=1S/C13H12N4O/c1-17-11-6-9(7-15-12(11)16-13(17)14)8-2-4-10(18)5-3-8/h2-7,18H,1H3,(H2,14,15,16)
Formel vikt 240.1011