Läs mer
4-Hydroxy Toremifene, TRC
Produktkod. 30004849
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004849 10 mg
30010798 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004849 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95450010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110503-62-3
Molekylformel C26 H28 Cl N O2
Molekylvikt (g/mol) 421.96
Synonym Phenol, 4-[(1Z)-4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]- (ACI), 4-[(1Z)-4-Chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenol (ACI), Phenol, 4-[(1Z)-4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]- (9CI), Phenol, 4-[4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]-, (Z)- (ZCI), (Z)-4-Hydroxytoremifene, 4-Hydroxytoremifene, 4-Hydroxy Toremifene
IUPAC-namn (Z)-4-(4-chlorophenyl)-4-[4-[2-(dimethylamino)ethoxy]phenyl]-3-phenylbut-3-en-1-ol
LEDER CN(C)CCOc1ccc(cc1)\C(=C(/CCO)\c2ccccc2)\c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material 4-Hydroxy Toremifene
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C26H28ClNO2/c1-28(2)17-19-30-24-14-10-22(11-15-24)26(21-8-12-23(27)13-9-21)25(16-18-29)20-6-4-3-5-7-20/h3-15,29H,16-19H2,1-2H3/b26-25+
Formel vikt 421.1809
Procent renhet >95%