Läs mer
4-Hydroxy Trimethoprim, TRC
Produktkod. 30133236
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133236 50 mg
30006183 10 mg
30006287 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133236 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH97198050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112678-48-5
Molekylformel C14H18N4O4
Molekylvikt (g/mol) 306.32
Synonym 2,6-Diamino-5-[(3,4,5-trimethoxyphenyl)methyl]-4(5H)-pyrimidinone
LEDER COC1=CC(=CC(=C1OC)OC)CC2=C(NC(=NC2=O)N)N
Specifikationer
Kemiskt namn eller material 4-Hydroxy Trimethoprim
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C14H18N4O4/c1-20-9-5-7(6-10(21-2)11(9)22-3)4-8-12(15)17-14(16)18-13(8)19/h5-6H,4H2,1-3H3,(H5,15,16,17,18,19)
Formel vikt 306.1328
Procent renhet >95%