Learn More
4′-Hydroxyacetophenone, Sigma-Aldrich™
Product Code. 15829120
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829120 250 g 250g
15819120 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15829120 Supplier Sigma Aldrich Supplier No. 8043110250
 Restricted Item
Beräknad leverans: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-93-4
Grade Research
Molecular Formula 4-(OH)C6H4COCH3
Molecular Weight (g/mol) 136.15
Specifications
Melting Point 105°C to 108°C
MDL Number MFCD00002359
Quantity 250 g
Synonym 4′-Hydroxyacetophenone, 4-Acetylphenol, 4′-Hydroxyacetophenone