Läs mer
4-Hydroxybenzoic-1,2,3,4,5,6-13C6 Acid, TRC
Produktkod. 30135277
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135277 100 mg
30010959 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135277 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH808453100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 287399-29-5
Molekylformel 13C6 C H6 O3
Molekylvikt (g/mol) 144.08
Synonym Benzoic-1,2,3,4,5,6-13C6 acid, 4-hydroxy- (9CI), p-Salicylic acid-13C6 (phenyl-13C6), 4-Carboxyphenol-13C6, 4-Hydroxybenzene-13C6-carboxylic acid, p-Carboxyphenol-13C6, p-Hydroxybenzoic acid-13C6, 4-Hydroxy-1,2,3,4,5,6-13C6-benzoic acid
IUPAC-namn 4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-carboxylic acid
LEDER O[13c]1[13cH][13cH][13c]([13cH][13cH]1)C(=O)O
Specifikationer
Kemiskt namn eller material 4-Hydroxybenzoic acid phenyl-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 144.0518
Procent renhet >95%