Learn More
4-Hydroxybenzoic acid-propyl ester sodium, Dr. Ehrenstorfer
Product Code. 30317947
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
30317947 250 mg 250mg
1 options
Product Code. 30317947 Supplier Dr. Ehrenstorfer Supplier No. DREC14229230
 Restricted Item
Beräknad leverans: 27-11-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium Propyl Parahydroxybenzoate
CAS 35285-69-9
Analyte or Component Names Sodium Propyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C10 H11 O3 . Na
Certifications/Compliance ISO 17034
Synonym Sodium Propyl Parahydroxybenzoate
InChI Formula InChI=1S/C10H12O3.Na/c1-2-7-13-10(12)8-3-5-9(11)6-4-8;/h3-6,11H,2,7H2,1H3;/q;+1/p-1
Solution Type Neat
SMILES [Na+].CCCOC(=O)c1ccc([O-])cc1
IUPAC Name sodium;4-propoxycarbonylphenolate
Industry Type Environmental Testing
Molecular Weight (g/mol) 202.18
Quantity 250 mg
Formula Weight 202.0606 g/mol