Läs mer
4-Hydroxyestradiol 3-O-b-D-Glucuronide, TRC
Produktkod. 30408290
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30408290 5 mg
1 options
Produktkod. 30408290 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9416055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxyestradiol 3-O-beta-D-Glucuronide
CAS 90746-94-4
Molekylformel C24H32O9
Kvantitet 5 mg
Synonym (17beta)-4,17-Dihydroxyestra-1,3,5(10)-trien-3-yl beta-D-Glucopyranosiduronic Acid, 4-Hydroxyestradiol 3-Glucuronide, Estrane beta-D-Glucopyranosiduronic Acid derivative
InChI formel InChI=1S/C24H32O9/c1-24-9-8-11-10-4-6-15(32-23-20(29)18(27)19(28)21(33-23)22(30)31)17(26)13(10)3-2-12(11)14(24)5-7-16(24)25/h4,6,11-12,14,16,18-21,23,25-29H,2-3,5,7-9H2,1H3,(H,30,31)/t11-,12-,14+,16+,18+,19+,20-,21+,23-,24+/m1/s1
LEDER C[C@]12CC[C@H]3[C@@H](CCc4c(O)c(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)ccc34)[C@@H]1CC[C@@H]2O
IUPAC-namn (2S,3S,4S,5R,6S)-6-[[(8R,9S,13S,14S,17S)-4,17-dihydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 464.51
Formel vikt 464.2046