Läs mer
4-Hydroxyisovaleric Acid Sodium Salt, TRC
Produktkod. 30403599
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30403599 50 mg
1 options
Produktkod. 30403599 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH81289050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxyisovaleric Acid Sodium Salt
CAS 112895-77-9
Molekylformel C5H9NaO3
Kvantitet 50 mg
Synonym 4-Hydroxy-3-methyl-butanoic Acid Monosodium Salt
InChI formel InChI=1S/C5H10O3.Na/c1-4(3-6)2-5(7)8;/h4,6H,2-3H2,1H3,(H,7,8);/q;+1/p-1
LEDER [Na].O=C(O)CC(C)CO
Molekylvikt (g/mol) 140.11