Läs mer
4’-Hydroxyphenyl Carvedilol, TRC
Produktkod. 17930795
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17930795 50 mg
17942045 10 mg
17921395 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17930795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94912050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142227-49-4
Molekylformel C24H26N2O5
Molekylvikt (g/mol) 422.47
Synonym 4-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-3-methoxyphenol, 4-Hydroxycarvedilol, 4'-Hydroxyphenylcarvedilol, BM 140686, BM 14686, 4'-Hydroxyphenyl carvedilol
IUPAC-namn 4-[2-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-3-methoxyphenol
LEDER COc1cc(O)ccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23
Specifikationer
Kemiskt namn eller material 4'-Hydroxyphenyl Carvedilol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C24H26N2O5/c1-29-23-13-16(27)9-10-21(23)30-12-11-25-14-17(28)15-31-22-8-4-7-20-24(22)18-5-2-3-6-19(18)26-20/h2-10,13,17,25-28H,11-12,14-15H2,1H3
Formel vikt 422.1842
Procent renhet >95%