Läs mer
(4-Hydroxyphenyl)glycine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (4-Hydroxyphenyl)glycine |
| CAS | 938-97-6 |
| Molekylformel | C8 H9 N O3 |
| Kvantitet | 10 g |
| Synonym | 2-Amino-2-(4-hydroxyphenyl)acetic acid, Benzeneacetic acid, α-amino-4-hydroxy-, Glycine, 2-(p-hydroxyphenyl)- (6CI,7CI,8CI), α-Amino-4-hydroxybenzeneacetic acid, (RS)-2-(4-Hydroxyphenyl)glycine, (±)-2-(4-Hydroxyphenyl)glycine, (±)-2-(p-Hydroxyphenyl)glycine, (±)-Amino(p-hydroxyphenyl)acetic acid, (±)-p-Hydroxyphenylglycine, (±)-α-Amino-4-hydroxybenzeneacetic acid, 2-(4-Hydroxyphenyl)glycine, 2-Amino-2-(4-hydroxyphenyl)aceticacid, DL-2-(4-Hydroxyphenyl)glycine, DL-2-(p-Hydroxyphenyl)glycine, DL-4-Hydroxyphenylglycine, DL-p-Hydroxyphenylglycine, DL-α-Amino(4-hydroxyphenyl)acetic acid, NSC 30081, p-Hydroxyphenylglycine, α-(4-Hydroxyphenyl)glycine, α-(p-Hydroxyphenyl)glycine, α-Amino-4-hydroxyphenylacetic acid, α-Amino-p-hydroxyphenylacetic acid |
| InChI formel | InChI=1S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10H,9H2,(H,11,12) |
| LEDER | NC(C(=O)O)c1ccc(O)cc1 |
| IUPAC-namn | 2-amino-2-(4-hydroxyphenyl)acetic acid |
| Molekylvikt (g/mol) | 167.162 |
| Formel vikt | 167.0582 |