Läs mer
(±)-4-Hydroxypropranolol HCl, CDN
Produktkod. 30321938
Change view
Kvantitet:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30321938 0.05 g
30321415 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30321938 Leverantör CDN Leverantörsnummer CDNX43290.05G
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+/-)-4-Hydroxy Propranolol Hydrochloride
CAS 14133-90-5
Molekylformel C16 H21 N O3 . Cl H
Kvantitet 0.05 g
Synonym 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (9 CI), 2-Propanol, 1-[(4-hydroxy-1-naphthyl)oxy]-3-(isopropylamino)-, hydrochloride (8 CI), 4-Hydroxypropranolol hydrochloride, 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (1:1), 4-Hydroxypropranolol Hydrochloride
InChI formel InChI=1 S/C16H21NO3.ClH/c1-11(2)17-9-12(18)10-20-16-8-7-15(19)13-5-3-4-6-14(13)16;/h3-8,11-12,17-19 H,9-10H2,1-2H3;1 H
LEDER Cl.CC(C)NCC(O)COc1ccc(O)c2ccccc12
IUPAC-namn 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]naphthalen-1-ol;hydrochloride
Molekylvikt (g/mol) 311.8038
Formel vikt 311.1288 g/mol
Procent renhet min 98% chemical purity