missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog Number p-7022887
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
611-36-9
C9H7NO
145.16
MFCD00006777,MFCD00956391
PMZDQRJGMBOQBF-UHFFFAOYSA-N
4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
69141
CHEBI:15815
O=C1C=CNC2=CC=CC=C12
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
611-36-9
C9H7NO
145.16
MFCD00006777,MFCD00956391
PMZDQRJGMBOQBF-UHFFFAOYSA-N
4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
69141
CHEBI:15815
O=C1C=CNC2=CC=CC=C12

4-Hydroxyquinoline is a medical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyquinoline is a medical raw material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY
Melting Point 201°C to 205°C
Quantity 1 g
Beilstein 1524969
Solubility Information Soluble in methanol.
IUPAC Name 1H-quinolin-4-one
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'