Läs mer
4-Hydroxytryptamine Creatinine Sulfate, TRC
Produktkod. 30011253
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011253 5 mg
30135516 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011253 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9748805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55206-11-6
Molekylformel C10 H12 N2 O . C4 H10 N4 O . H2 O4 S
Molekylvikt (g/mol) 404.44
Synonym 4-Hydroxytryptamine Creatinine
IUPAC-namn 3-(2-aminoethyl)-1H-indol-4-ol;2-[carbamimidoyl(methyl)amino]acetamide;sulfuric acid
LEDER CN(CC(=O)N)C(=N)N.NCCc1c[nH]c2cccc(O)c12.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material 4-Hydroxytryptamine Creatinine Sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C10H12N2O.C4H10N4O.H2O4S/c11-5-4-7-6-12-8-2-1-3-9(13)10(7)8;1-8(4(6)7)2-3(5)9;1-5(2,3)4/h1-3,6,12-13H,4-5,11H2;2H2,1H3,(H2,5,9)(H3,6,7);(H2,1,2,3,4)
Formel vikt 404.1478
Procent renhet >95%