Läs mer
4'-(Imidazol-1-yl)acetophenone, TRC
Produktkod. 30395653
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30395653 1 g
30395974 10 g
30406620 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30395653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI5774631G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 10041-06-2
Molekylformel C11H10N2O
Molekylvikt (g/mol) 186.21
IUPAC-namn 1-(4-imidazol-1-ylphenyl)ethanone
LEDER CC(=O)c1ccc(cc1)n2ccnc2
Specifikationer
Kemiskt namn eller material 4'-(Imidazol-1-yl)acetophenone
Kvantitet 1 g
InChI formel InChI=1S/C11H10N2O/c1-9(14)10-2-4-11(5-3-10)13-7-6-12-8-13/h2-8H,1H3
Formel vikt 186.0793