Learn More
4-Iodoaniline, 99%, Thermo Scientific Chemicals
Catalog Number p-4510315
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
540-37-4
C6H6IN
219.02
VLVCDUSVTXIWGW-UHFFFAOYSA-N
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
10893
4-iodoaniline
C1=CC(=CC=C1N)I
540-37-4
C6H6IN
219.02
VLVCDUSVTXIWGW-UHFFFAOYSA-N
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
10893
4-iodoaniline
C1=CC(=CC=C1N)I
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 59.0°C to 64.0°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4NH2
Quantity 25 g
Beilstein 12,67
Merck Index 15,507
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol,chloroform and ether
Formula Weight 219.02
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4-Iodoaniline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodoaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.