missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is used as intermediate in organic syntheses & in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as intermediate in organic syntheses & in medicine.
Solubility
Reacts with water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
It is used as intermediate in organic syntheses & in medicine.
Solubility
Reacts with water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
Specifications
Melting Point | 79°C to 84°C |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 2691661 |
Sensitivity | Light and moisture sensitive |
Merck Index | 14,7486 |
Solubility Information | Reacts with water. |
Formula Weight | 302.52 |
Percent Purity | 97% |
Chemical Name or Material | 4-Iodobenzenesulfonyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'