missing translation for 'onlineSavingsMsg'
Learn More

4-Iodopyridine, 96%, Thermo Scientific Chemicals

Catalog Number p-7028251
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15854-87-2
C5H4IN
205.00
MFCD02093937
RTLUPHDWSUGAOS-UHFFFAOYSA-N
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
609492
4-iodopyridine
IC1=CC=NC=C1
This item is not returnable. View return policy
15854-87-2
C5H4IN
205.00
MFCD02093937
RTLUPHDWSUGAOS-UHFFFAOYSA-N
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
609492
4-iodopyridine
IC1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 96°C to 99°C
Quantity 1 g
Beilstein 1594
Sensitivity Light sensitive
Formula Weight 205
Percent Purity 96%
Chemical Name or Material 4-Iodopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'