Läs mer
4-Keto 13-cis-Retinoic Acid, TRC
Produktkod. 30003691
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30003691 10 mg -
30010339 25 mg -
30006243 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK20497010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71748-58-8
Molekylformel C20H26O3
Molekylvikt (g/mol) 314.42
Synonym BAL10085, BAL0010085-000, Isotretinoin EP Impurity H, BAL0004079-000, BAL4079, 9-cis retinoic acid, alitretinoin related, 13-cis-4-oxo retinoic acid
IUPAC-namn (2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
LEDER C\C(=C\C(=O)O)\C=C\C=C(/C)\C=C\C1=C(C)C(=O)CCC1(C)C
Specifikationer
Kemiskt namn eller material 4-Keto 13-cis-Retinoic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13-
Formel vikt 314.1882