missing translation for 'onlineSavingsMsg'
Läs mer

4-Methoxy-2,3,6-trimethylbenzaldehyde-d3, TRC

Produktkod. 30479888
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479888 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30479888 Supplier Toronto Research Chemicals Supplier No. TRCM269702250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Methoxy-2,3,6-trimethylbenzaldehyde-d3
CAS 1216683-89-4
Molekylformel C11 2H3 H11 O2
Kvantitet 250 mg
Synonym 2,3,6-Trimethyl-4-(trideuteriomethoxy)benzaldehyde, 4-Methoxy-2,3,6-trimethylbenzaldehyde-d3, 2,3,6-Trimethyl-p-anisaldehyde-d3, 4-Methoxy-2,3,6-trimethylbenzaldehyde-D3 (methoxy-D3)
InChI formel InChI=1S/C11H14O2/c1-7-5-11(13-4)9(3)8(2)10(7)6-12/h5-6H,1-4H3/i4D3
LEDER [2H]C([2H])([2H])Oc1cc(C)c(C=O)c(C)c1C
IUPAC-namn 2,3,6-trimethyl-4-(trideuteriomethoxy)benzaldehyde
Molekylvikt (g/mol) 181.25
Formel vikt 181.1182
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.